MycoLight™ Bacterial Viability Assay Kit
Catalog # AATB22400
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pk:200 Tests
- Description:MycoLight™ bacterial viability assay kit
- Environmentally Preferable:
- Tests per kit:200 Tests
Specifications
About this item
Mycolight™ Bacterial Viability Assay Kit provides two-colour fluorescence assay of bacterial viability in both gram-positive and negative bacterial cell.
- Suitable for gram-positive and gram-negative bacteria
- Contains all necessary components
- Multiple applications
Caution: For research use only.