Order Entry
Denmark
ContactUsLinkComponent
Screen Quest™ Fluorimetric Fatty Acid Uptake Assay Kit
Screen Quest™ Fluorimetric Fatty Acid Uptake Assay Kit
Catalog # AATB36385
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Screen Quest™ Fluorimetric Fatty Acid Uptake Assay Kit
Catalog # AATB36385
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    100 Tests
  • Description:
    Screen Quest™ Fluorimetric fatty acid uptake assay kit
  • Tests per kit:
    100 Tests

Specifications

About this item

Fatty acid uptake is an important therapeutic target for the treatment of many human diseases such as obesity, type 2 diabete and hepatic steatosis.

  • Improved sensitivity
  • Contains all necessary components
  • Robust and adaptable protocol
  • Multiple applications
Caution: For research use only.