PhosphoWorks™ Luminometric ATP Assay Kit, Steady Glow
Leverandør: AAT BIOQUEST
Certifikater
Om denne vare
Adenosine triphosphate (ATP) plays a fundamental role in cellular energenics, metabolic regulation and cellular signaling.
- Extreme sensitivity
- Stable signal with >4 hour half-life
- No mixing or separations required
Caution: For research use only.
Frequently Bought Together