Order Entry
Denmark
ContactUsLinkComponent
Cell Meter™ Fluorimetric Intracellular Total ROS Activity Assay Kit*Red Fluorescence*
Cell Meter™ Fluorimetric Intracellular Total ROS Activity Assay Kit*Red Fluorescence*
Catalog # AATB22901
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Cell Meter™ Fluorimetric Intracellular Total ROS Activity Assay Kit*Red Fluorescence*
Catalog # AATB22901
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    200 Tests
  • Description:
    Cell Meter™ fluorimetric Intracellular total ROS activity assay kit *red fluorescence*
  • Tests per kit:
    200 Tests

Specifications

About this item

Reactive oxygen species (ROS) are natural byproducts of the normal metabolism of oxygen and play important roles in cell signaling.

  • Ready to use
  • Contains all necessary components
  • Robust and adaptable
  • Suitable for all major fluorescence platforms
  • No separation step needed
Caution: For research use only.