Order Entry
Denmark
ContactUsLinkComponent
MitoLite™ Orange EX405
MitoLite™ Orange EX405
Catalog # AATB22679
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
MitoLite™ Orange EX405
Catalog # AATB22679
Supplier:  AAT BIOQUEST
CAS Number:  
Specifications
  • Pk:
    500 Tests
  • Description:
    MitoLite™ Orange 405
  • Tests per kit:
    500 Tests
Specifications
About this item
MitoLite™ Orange 405 is a cationic dye that selectively accumulates in mitochondria probably vial the mitochondrial membrane potential gradient.
  • Mitochondria-Specific Dye
  • Cell-Permeable
  • Robust
Caution: For research use only.