Cell Meter™ Fixable Viability Dye, BX650
Catalog # AATB22520
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pk:200 Tests
- Description:Cell Meter™ BX650 fixable cell stain
- Purity:90%
- Tests per kit:200 Tests
Specifications
About this item
Discrimination and exclusion of dead cells from live cells allows cleaner separation and identification of cell populations.
- Cell-Impermeable
- Amine- And Thiol-Reactive
- High Stability
- Live/Dead Cell Labeling Dye
- Fixable
Caution: For research use only.