Order Entry
Denmark
ContactUsLinkComponent
CytoTrace™ Red CFDA
CytoTrace™ Red CFDA
  AATB22016
 :  
undefined
CytoTrace™ Red CFDA
  AATB22016
 :  
  • Pk:
    1 mg
  • Description:
    CytoTrace™ Red CFDA
  • Purity:
    90%
CytoTrace™ Red fluorescent probe retains the spectral properties of Cy3/TRITC, thus its signal can be easily monitored using fluorescence microscopy or flow cytometry.
  • High Multigenerational Cell Retention
  • Bright Signal
 : For research use only.