Order Entry
Denmark
ContactUsLinkComponent
Texas Red® Azide, Single Isomer
Texas Red® Azide, Single Isomer
Catalog # AATB484
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Texas Red® Azide, Single Isomer
Catalog # AATB484
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Description:
    Texas Red® azide
  • Purity:
    95%

Specifications

About this item

Texas Red® azide is an excellent alkyne-reactive fluorescent reagent for labeling alkyne-containing biological molecules through the well known click chemistry.

  • Identical spectra to Texas Red
  • High Stability
Caution: For research use only.