Texas Red® Cadaverine, Single Isomer
Catalog # AATB482
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pk:5 mg
- Description:Texas Red® cadaverine
- Purity:95%
Specifications
About this item
Texas Red® cadaverine reacts with carbonyl compounds such as aldehyde, ketones and acids to give bright red fluorescent conjugates.
- Identical spectra to Texas Red
- High Stability
Caution: For research use only.