Order Entry
Denmark
Orders LinkContactUsLinkComponent
RS-232 Cable
RS-232 Cable
RS-232 cable, 25-pin – 9-pin, RS-232, 3 m
Catalog # METR6.2125.110
Supplier:   METROHM
RS-232 Cable
Catalog # METR6.2125.110
Supplier:  METROHM
RS-232 Cable
RS-232 cable, 25-pin – 9-pin, RS-232, 3 m
Price
980,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Connecting RS-232 interface to PC. Used with pH meters, titrinos, KF coulometers, VA trace analyser.
Specifications
  • Type:
    RS-232
  • Description:
    RS-232 cable, 25-pin – 9-pin
  • Length:
    3 m