
Biotin maleimide, ReadiView™ biodetection
Catalog # AATB3058
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:451.54 g/mol
ADR:5,II
:GHS07
CAS Number:116919-18-7
Formula:C20H29N5O5S