0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Denmark
ContactUsLinkComponent
PerCP
PerCP, ReadiUse™ fluorescent probe, preactivated
Irriterende
PerCP, ReadiUse™ fluorescent probe, preactivated
Catalog # AATB2590
Leverandør:
AAT BIOQUEST
CAS nummer:
Expand image
Irriterende
PerCP, ReadiUse™ fluorescent probe, preactivated
Catalog # AATB2590
Leverandør:
AAT BIOQUEST
CAS nummer:
Specifikationer
Pack type:
Plastic bottle
Pk:
1 mg
Seq:
2070
Specifikationer
Om denne vare
Product Details
:
:
350000 g/mol
ADR:
5,II
:
GHS07
Product Details