Order Entry
Denmark
ContactUsLinkComponent
 
Irriterende
PE-Cy5 Tandem, ReadiUse™ fluorescent probe, preactivated
Catalog # AATB2580
Leverandør:  AAT BIOQUEST
CAS nummer:  
undefined
Irriterende
PE-Cy5 Tandem, ReadiUse™ fluorescent probe, preactivated
Catalog # AATB2580
Leverandør:  AAT BIOQUEST
CAS nummer:  

Specifikationer

  • Pack type:
    Plastic bottle
  • Pk:
    1 mg
  • Seq:
    2070

Specifikationer

Om denne vare

 :
 :240000 g/mol
ADR:5,II
 :GHS07