
Trypan blue 10 mM live cell labeling
Catalog # AATB2453
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:10 mL
- Seq:2275
Specifications
About this item
:
:960.82 g/mol
ADR:5,II
:GHS07
CAS Number:72-57-1
Formula:C₃₄H₂₄N₆Na₄O₁₄S₄
MDL Number:MFCD00003969