Order Entry
Denmark
ContactUsLinkComponent
Tide Fluor™ 1 succinimidyl ester, Tide Fluor™ fluorescent probe
Irritant
Tide Fluor™ 1 succinimidyl ester, Tide Fluor™ fluorescent probe
Catalog # AATB2244
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Irritant
Tide Fluor™ 1 succinimidyl ester, Tide Fluor™ fluorescent probe
Catalog # AATB2244
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :330.29 g/mol
ADR:5,II
 :GHS07