Order Entry
Denmark
Orders LinkContactUsLinkComponent
(Ac-KQL)2R110

(Ac-KQL)2R110

Plastic bottle, 1 mg
Catalog # AATB13465
Irritant
Supplier:   AAT Bioquest
(Ac-KQL)2R110
Catalog # AATB13465
Supplier:  AAT Bioquest
CAS Number:  
(Ac-KQL)2R110
(Ac-KQL)2R110

Plastic bottle, 1 mg

CAS Number:  
Price
1.170,00 kr
Log in for availability
1 * 1 mg
Product Details & Documents

The non-fluorescent R110 substrates generate the bright green fluorescent rhodamine 110 product that has Ex/Em = 494/521 nm, and can be easily detected with a FITC filter set.

  • Protease substrate
  • Bright signal
  • Multiple applications
  • ADR:5,II
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 mg

Specifications

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.