0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Denmark
ContactUsLinkComponent
Violet 540 acid
Violet 540 acid, mFluor™ fluorescent probe
Violet 540 acid, mFluor™ fluorescent probe
AATB1142
:
AAT BIOQUEST
:
Violet 540 acid, mFluor™ fluorescent probe
AATB1142
:
AAT BIOQUEST
:
Pack type:
Plastic bottle
Pk:
5 mg
Seq:
2210
:
:
1600 kDa
:
5,II
:
GHS07