iFluor® 488, Hydrazide
Plastic bottle, 1 mg
Catalog # AATB1082
Supplier: AAT Bioquest
Gå til:
Product Details & Documents
Although FITC is still the most popular fluorescent labeling dye for preparing green fluorescent bioconjugates, there are certain limitations with FITC, such as severe photobleaching for microscope imaging and pH-sensitive fluorescence.
- Bright and photostable
- Minimal quenching
- Stable conjugation
- pH independent fluorescence between 4-10
- ADR:5,II
Specifikationer
Specifikationer
- Pack type:Plastic bottle
- Pk:1 mg



