Order Entry
Denmark
ContactUsLinkComponent
SPINeasy® RNA Extraction Kit for Virus, MP Biomedicals
SPINeasy® RNA Extraction Kit for Virus, MP Biomedicals
Catalog # 116537050.
Supplier:  MP Biomedicals
CAS Number:  
undefined
SPINeasy® RNA Extraction Kit for Virus, MP Biomedicals
Catalog # 116537050.
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Method / Format:
    Column (silica-based)
  • Sample type:
    Saliva
    Serum
  • Target molecule:
    Viral RNA
  • Description:
    SPINeasy® RNA extraction kit for virus, 50 tests
  • Storage temperature:
    Store at room temperature (15...30 °C), in a dry environment.

Specifications

About this item

SPINeasy Virus RNA kit is a silica-membrane spin-column kit that enables quick and convenient extraction of virus RNA from cell culture media and bodily fluids such as saliva and serum, without the use of toxic substances such as phenol and chloroform. Through a simple workflow, virus RNA is typically extracted within 20 minutes and is immediately available for downstream applications such as RT-PCR and RT-qPCR.

The SPINeasy virus RNA kit quickly and efficiently isolates high-quality, total RNA from cell culture media and bodily fluids in approximately 15 minutes using and SPIN columns for the purification process without any toxic chemicals.

RNA extracted from SPINeasy virus RNA kit can be used in several downstream applications, including RT-PCR, qPCR, RNA-Seq,, and northern blot analysis, genomics, molecular biology.

Delivery: Kit includes 30 ml Lysis Buffer VR, 6 ml Wash Buffer VR, 10 ml Nuclease-free water, 1 vial Carrier RNA, 50× Column VR with collection tubes.