Order Entry
Denmark
Orders LinkContactUsLinkComponent
Color mix, complementary batch
Complementary kit for color mixing
  765-1314
Marketsource
 :   PHYWE
Color mix, complementary batch
  765-1314
 :  PHYWE
Marketsource
Color mix, complementary batch
Color mix, complementary batch
Complementary kit for color mixing
1 * 1 Items
Supplementary kit for table optics 765-1317. Content:
  • 2 mirrors
  • 1 column
  • 3 filters for additive color mixing
  • 3 filters for subtractive color mixing
  • Description:
    Complementary kit for color mixing