Order Entry
Denmark
Orders LinkContactUsLinkComponent
Picture optics, completion kit
Picture optics, completion kit
Supplementary kit with lenses
Catalog # 765-1009
Supplier:   KVANT LIFE SCIENCE
Picture optics, completion kit
Catalog # 765-1009
Supplier:  KVANT LIFE SCIENCE
Picture optics, completion kit
Picture optics, completion kit
Supplementary kit with lenses
Price
Inquire For Price
Log in for availability
1 * 1 Each
Product Details & Documents
New optical models for laser optics (15 parts). Consists of.
  • concave lenses
  • plank concave lenses
  • convex lenses
  • planar convex lenses
  • equilateral prism
  • rectangular prisms
  • mirrors
  • condenser lenses
  • cubic beam splitters
Specifications
  • Description:
    Supplementary kit with lenses