Spectrum lamps with Pico 9 base
Leverandør: OSRAM
Certifikater
Om denne vare
For the production of monochromatic light together with suitable filters or for line spectra experimentation. The spectral lamps may only be connected to 220V AC via the choke.
Operating current: 1 A
Burn mode: Vertically standing
Socket: Pico 9
Frequently Bought Together