Thermocouple
Thermocouple
Catalog # 765-0212
Marketsource
Supplier: CORNELSEN EXPERIMENTA
Jump to:
Product Details & Documents
For experiments with thermoelectric effects. Consists of brazed copper and constant wire. Equipped with connections for 4 mm banana connectors. Mounted on plate with rod, 10 mm.
Specifications
- Description:Thermocouple