Engine models
Marketsource
Supplier: CORNELSEN EXPERIMENTA
Jump to:
About this item
Intended for use on overhead projectors for demonstration of function and principle for different types of motors. Made of strong, transparent plastic. Equipped with suction feet.
- Dimensions: 250 × 250 mm
5 Options Available Below
Product Details & Documents
Intended for use on overhead projectors for demonstration of function and principle for different types of motors. Made of strong, transparent plastic. Equipped with suction feet.
- Dimensions: 250 × 250 mm
Product Family Options
Product Information
- AvailabilityPrice
- 765-0176Diesel engine, four-strokeSpecifications:MarketsourceMarketsource
- 765-0175Otto engine, four-strokeSpecifications:MarketsourceMarketsource
- 765-0174Otto engine, two-strokeOtto engine, two-strokeDiscontinued-
- 765-0173Wobbling motorSpecifications:MarketsourceMarketsource
- 765-0172Stirling engineSpecifications:MarketsourceMarketsource
- 765-0171Steam engineSpecifications:MarketsourceMarketsource