- Antibody type:Secondary
- Antigen name:IGHM
- Clonality:Polyclonal
- Host:Goat
- Isotype:IgG
- Reactivity:Rat
- Gene ID:3507
- Purity:Multi-step
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2
- Storage temperature:Store at 4 °C short term. Aliquot and store at −20 °C long term. Avoid freeze-thaw cycles.
- Immunogen:Rat IgM whole molecule
- Pk:1 mg
This product was prepared from monospecific antiserum by immunoaffinity chromatography using Rat IgM coupled to agarose beads followed by solid phase adsorption(s) to remove any unwanted reactivities. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Goat Serum, Rat IgM and Rat serum.
Immunohistochemistry: 1:1000 - 1:5000
Immunohistochemistry-Paraffin: 1:1000 - 1:5000
Western Blot: 1:2000 - 1:10000
This product has been tested by ELISA and western blot and is assayed against Rat IgM in a standard capture ELISA using Peroxidase Conjugated Streptavidin. A working dilution of 1:15,000 to 1:70,000 of the concentration is suggested for this product. Rat IgM antibody is suitable for immunoassays including western blot and ELISA.
Type: Secondary
Antigen: IGHM
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rat