Anti-ZEB2 Rabbit Polyclonal Antibody
NOVUNBP1-82991
- Antibody type:Primary
- Antigen name:ZEB2 (Zinc finger E-box binding homeobox 2)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Gene ID:9839
- Antigen synonyms:Zinc finger homeobox protein 1b|zinc finger E-box-binding homeobox 2|zinc finger E-box binding homeobox 2|SIP1HSPC082|HSPC082|Smad-interacting protein 1|SIP1|SMADIP1ZFHX1BSIP-1|ZFX1B|SMADIP1|ZFHX1B|KIAA0569FLJ42816|ZEB2|zinc finger homeobox 1b
- Purity:Immunogen affinity purified
- Storage buffer:PBS (pH 7,2) and 40% Glycerol
- Storage temperature:Store at 4 °C short term. Aliquot and store at −20 °C long term. Avoid freeze-thaw cycles.
- Immunogen:This antibody was developed against Recombinant Protein corresponding to amino acids: SPVKPMDSITSPSIAELHNSVTNCDPPLRLTKPSHFTNIKPVEKLDHSRSNTPSPLNLSSTSSKNSHSSSYTPNSFSSEELQAEPLDLSLPKQMKEPKSIIATKNKTKASSISLDHNSVSSSSE
- Pk:0,1 mL
Analysis in human cell lines SK-MEL-30 and Caco-2 using Anti-ZEB2 antibody. Corresponding ZEB2 RNA-seq data are presented for the same cell lines. Loading control: Anti-HDAC1.
- Immunohistochemistry 1:500 - 1:1000
- Immunohistochemistry - Paraffin: 1:500 - 1:1000
- Western blot: 0.04 to 0.4 ug/ml
Type: Primary
Antigen: ZEB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse