Order Entry
Denmark
ContactUsLinkComponent
VWR® Short Thread Vials ND9
VWR® Short Thread Vials ND9
Catalog # 548-2644A
Supplier:  VWR Collection
VWR® Short Thread Vials ND9
Catalog # 548-2644A
Supplier:  VWR Collection
Specifications
  • Ø×H:
    11,6×32,0 mm
  • Packed:
    100 per pack
  • Capacity:
    0,20 ml
  • Colour:
    Clear
  • Description:
    Short thread vial with integrated micro-insert
  • Material:
    Glass
  • Pk:
    1.000
Specifications
About this item
The vials can be used on all common autosamplers due to their technical geometry.
  • 1st hydrol. class, label and filling lines
  • Top bonded