Order Entry
Denmark
ContactUsLinkComponent
Potassium hydroxide 0.2 mol/l (0.2 N)
Potassium hydroxide 0.2 mol/l (0.2 N)
Catalog # ACSDZ14574519.1L
Supplier:  CPACHEM
CAS Number:  
Potassium hydroxide 0.2 mol/l (0.2 N)
Catalog # ACSDZ14574519.1L
Supplier:  CPACHEM
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :56.11 g/mol
 :GHS05
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553