Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Denmark
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Media for Microbiology
Dehydrated Media for Microbiology
Luria broth media
Luria broth media
Luria broth media
Catalog # EDVO611
Supplier:
EDVOTEK
CAS Number:
Expand image
Luria broth media
Catalog # EDVO611
Supplier:
EDVOTEK
CAS Number:
Specifications
Pk:
100 g
Standards:
-
Description:
LB media
Specifications
About this item
Product Details
Product Details