Methoxy-PEG ELISA kit
Catalog # ABNOKA6893.1KIT
Supplier: Abnova
CAS Number:
Specifications
- Pk:1 KIT
- Target protein:Methoxy-PEG
Specifications
About this item
Methoxy-PEG ELISA Kit is a sandwich-type ELISA and use two different monoclonal antibodies. The capture antibody recognizes the polyoxyethylene backbone. The other is specific for the terminal methoxy group of mPEGs and mPEGylated proteins.