Treponema pallidum IgG ELISA kit
Catalog # ABNOKA6891.1KIT
Supplier: Abnova
CAS Number:
Specifications
- Pk:1 KIT
- Primary antibody reactivity:Treponema pallidum
- Target protein:IgG
- Assay Principle:Quantitative
Specifications
About this item
Treponema pallidum IgG ELISA Kit is used for the qualitative and semiquantitative detection of IgG antibody to Treponema pallidum in serum or plasma.