Order Entry
Denmark
ContactUsLinkComponent
 
The CA 15-3 ELISA kit
  ABNOKA6887.1KIT
 :  Abnova
 :  
The CA 15-3 ELISA kit
  ABNOKA6887.1KIT
 :  Abnova
 :  

 

  • Pk:
    1 KIT
  • Target protein:
    CA 15-3
  • Assay Principle:
    Quantitative

 

 

The CA 15-3 ELISA Kit is an enzyme immunoassay for the quantitative in vitro diagnostic measurement of CA 15-3 in serum and plasma.