Ovalbumin Specific IgE (Human) ELISA kit
Catalog # ABNOKA6788.1KIT
Supplier: Abnova
CAS Number:
Specifications
- Pk:1 KIT
- Primary antibody reactivity:Human
- Target protein:Ovalbumin Specific IgE
- Assay Principle:Qualitative
Specifications
About this item
Ovalbumin Specific IgE (Human) ELISA Kit is for the qualitative determination of human ovalbumin specific IgE concentrations in serum or plasma.