The IgG (Monkey) ELISA kit
Catalog # ABNOKA6754.1KIT
Supplier: Abnova
CAS Number:
Specifications
- Pk:1 KIT
- Primary antibody reactivity:Monkey
- Target protein:IgG
Specifications
About this item
The IgG (Monkey) ELISA Kit is used for immunoperoxidase Assay for Determination of IgG in Monkey Samples.