IgA (Sheep) ELISA kit
Catalog # ABNOKA6764.1KIT
Supplier: Abnova
CAS Number:
Specifications
- Pakn:1 KIT
- Primary antibody reactivity:Sheep
- Target protein:IgA
- Assay princip:Quantitative
Specifications
About this item
IgA (Sheep) ELISA Kit is for the quantitative measurement of sheep IgA in serum or plasma.