DNP IgG (Mouse) ELISA kit
Catalog # ABNOKA6739.1KIT
Supplier: Abnova
CAS Number:
Specifications
- Pk:1 KIT
- Primary antibody reactivity:Mouse
- Target protein:IgG
- Assay Principle:Quantitative
Specifications
About this item
DNP IgG (Mouse) ELISA Kit is for the quantitative measurement of mouse anti-DNP IgG in serum or plasma samples.