Order Entry
Denmark
ContactUsLinkComponent
Brucella IgM ELISA kit
Brucella IgM ELISA kit
Catalog # ABNOKA6721.1KIT
Supplier:  Abnova
CAS Number:  
Brucella IgM ELISA kit
Catalog # ABNOKA6721.1KIT
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Target protein:
    IgM
  • Assay Principle:
    Qualitative

Specifications

About this item

Brucella IgM ELISA Kit is a qualitative determination of IgM class antibodies against Brucella in human serum or plasma.