Order Entry
Denmark
ContactUsLinkComponent
Brucella IgG ELISA kit
Brucella IgG ELISA kit
Catalog # ABNOKA6720.1KIT
Supplier:  Abnova
CAS Number:  
Brucella IgG ELISA kit
Catalog # ABNOKA6720.1KIT
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Target protein:
    IgG
  • Assay Principle:
    Qualitative

Specifications

About this item

Brucella IgG ELISA Kit is a qualitative determination of IgG class antibodies against Brucella in human serum or plasma.