Adenovirus IgG (Human) ELISA kit
ABNOKA6714.1KIT
: Abnova
:
- Pk:1 KIT
- Primary antibody reactivity:Human
- Target protein:IgG
- Assay Principle:Qualitative
Adenovirus IgG (Human) ELISA Kit is a qualitative determination of IgG class antibodies against Adenovirus in human serum or plasma.