Order Entry
Denmark
ContactUsLinkComponent
Helicobacter pylori IgG ELISA kit
Helicobacter pylori IgG ELISA kit
Catalog # ABNOKA6696.1KIT
Supplier:  Abnova
CAS Number:  
Helicobacter pylori IgG ELISA kit
Catalog # ABNOKA6696.1KIT
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Primary antibody reactivity:
    H. pylori
  • Target protein:
    IgG
  • Assay Principle:
    Quantitative

Specifications

About this item

Helicobacter pylori IgG ELISA Kit is an enzyme immunoassay for the quantitative measurement of IgG antibodies to Helicobacter pylori in human serum and plasma.