Order Entry
Denmark
ContactUsLinkComponent
Prolactin (Human) ELISA kit
Prolactin (Human) ELISA kit
Catalog # ABNOKA6697.1KIT
Supplier:  Abnova
CAS Number:  
Prolactin (Human) ELISA kit
Catalog # ABNOKA6697.1KIT
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Primary antibody reactivity:
    Human
  • Target protein:
    Prolactin
  • Assay Principle:
    Quantitative

Specifications

About this item

Prolactin (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of prolactin in serum.