Order Entry
Denmark
ContactUsLinkComponent
Cytomegalovirus IgG ELISA kit
Cytomegalovirus IgG ELISA kit
Catalog # ABNOKA6698.1KIT
Supplier:  Abnova
CAS Number:  
Cytomegalovirus IgG ELISA kit
Catalog # ABNOKA6698.1KIT
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Primary antibody reactivity:
    Cytomegalovirus
  • Target protein:
    IgG
  • Assay Principle:
    Quantitative

Specifications

About this item

Cytomegalovirus IgG ELISA Kit is a sandwich enzyme immunoassay for the quantitative and qualitative measurement of Cytomegalovirus IgG in human serum and plasma.