Rubella IgG ELISA kit
Catalog # ABNOKA6701.1KIT
Supplier: Abnova
CAS Number:
Specifications
- Pk:1 KIT
- Primary antibody reactivity:Rubella virus
- Target protein:IgG
- Assay Principle:Quantitative
Specifications
About this item
Rubella IgG ELISA Kit is an enzyme immunoassay for the quantitative and qualitative measurement of IgG antibodies to Rubella Virus in human serum and plasma