CG (Human) ELISA kit
Catalog # ABNOKA6709.1KIT
Supplier: Abnova
CAS Number:
Specifications
- Pk:1 KIT
- Primary antibody reactivity:Human
- Target protein:CG
- Assay Principle:Quantitative
Specifications
About this item
CG (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of chorionic gonadotropin in serum and plasma.