Order Entry
Denmark
ContactUsLinkComponent
 
Synthetic Yeast Inorganic Pyrophosphatase (from E. coli)
  ABNOP9372.10ML
 :  Abnova
 :  
Synthetic Yeast Inorganic Pyrophosphatase (from E. coli)
  ABNOP9372.10ML
 :  Abnova
 :  

 

  • Pk:
    10 mL
  • Enzyme type:
    Synthetic
  • Source:
    E. coli
  • Species:
    Yeast
  • Enzyme name:
    Inorganic Pyrophosphatase