Order Entry
Denmark
Orders LinkContactUsLinkComponent
Recombinant Poly(A) Polymerase (from E. coli)

Recombinant Poly(A) Polymerase (from E. coli)

100 µl
Catalog # ABNOP9380.100UL
Supplier:   Abnova
Recombinant Poly(A) Polymerase (from E. coli)
Catalog # ABNOP9380.100UL
Supplier:  Abnova
CAS Number:  
Recombinant Poly(A) Polymerase (from E. coli)

100 µl

CAS Number:  
Price
13.880,00 kr
Log in for availability
1 * 100 µl
Product Details & Documents
Specifications

Specifications

  • Pk:
    100 µl
  • Cat. No.:
    ABNOP9380.100UL
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Enzyme name:
    Poly(A) Polymerase

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.