Recombinant Human TFF1 (from E. coli)
20 µG
Catalog # ABNOP9194.20UG
Supplier: Abnova
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:20 µG
- Cat. No.:ABNOP9194.20UG
- Protein/peptide type:Recombinant
- Protein/peptide name:TFF1
- Species:Human
- Source:E. coli



