Ordreindtastning
Denmark
ContactUsLinkComponent
 
Recombinant Human NENF (from E. coli), His-tag
Catalog # ABNOP8952.10UG
Supplier:  Abnova
CAS Number:  
Recombinant Human NENF (from E. coli), His-tag
Catalog # ABNOP8952.10UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pakn:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Kilde:
    E. coli
  • Species:
    Human
  • Tag sequence:
    His-tag
  • Gen ID:
    Q9UMX5
  • Protein/peptide name:
    NENF
  • Sekvens:
    32a.a.-172a.a.

Specifications

About this item