Recombinant Rat CXCL3 (from E. coli)
Catalog # ABNOP9473.10UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Rat
- Gene ID:Q10746
- Protein/peptide name:CXCL3
- Sequence:33a.a.-101a.a.