Recombinant Human CCL1 (from E. coli)
Catalog # ABNOP9478.10UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Gene ID:P22362
- Protein/peptide name:CCL1
- Sequence:23a.a.-96a.a.